Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Mandelic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-2-Hydroxy-2-phenylacetic acid DL-2-Hydroxy-2-phenylacetic acid DL-Mandelic acid

A14223-36 A14223-22 A14223-22 A14223-0E A14223-0E A14223-36
AAA14223-22EA 42.84 USD InStock
CAAAA14223-0E AAA14223-22 CAAAA14223-22 CAAAA14223-36 AAA14223-0E AAA14223-36
(±)-Mandelic acid 99%
(±)-Mandelic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 118…121 °C
Density: 1.361 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064250
CAS Number: 611-72-3
EINECS: 202-007-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty