Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-o-cresol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Hydroxybenzotrifluoride 2-(Trifluoromethyl)phenol

A14231-06 A14231-06 A14231-14 A14231-14 A14231-03 A14231-03
AAAA14231-03EA 44.72 USD InStock
CAAAA14231-14 AAAA14231-03 AAA14231-14 CAAAAA14231-03 CAAAA14231-06 AAA14231-06
α,α,α-Trifluoro-o-cresol 98%
α,α,α-Trifluoro-o-cresol


Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 147…148 °C
Melting Pt: 45…46 °C
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 1325
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty