Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2-chloroethyl)ether 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloroethyl ether

A14234-0B A14234-18 A14234-30 A14234-0B A14234-30 A14234-18
AAA14234-30EA 81 USD InStock
CAAAA14234-0B CAAAA14234-18 AAA14234-30 AAA14234-18 CAAAA14234-30 AAA14234-0B
Bis(2-chloroethyl)ether 99%
Bis(2-chloroethyl) ether


Formula: C₄H₈Cl₂O
MW: 143.01 g/mol
Boiling Pt: 178 °C
Melting Pt: –50 °C
Density: 1.219
Flash Pt: 55 °C(131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: "6.1",II
Merck Index: 13,03091
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty