Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14237-22 A14237-14 A14237-36 A14237-22 A14237-36 A14237-14
AAA14237-36EA 937.99 USD InStock
AAA14237-36 AAA14237-14 AAA14237-22 CAAAA14237-14 CAAAA14237-36 CAAAA14237-22
4-Bromobenzaldehyde 98+%
4-Bromobenzaldehyde


Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 66…68 °C/2 mm
Melting Pt: 55…58 °C
Density: 1,849 g/cm³ (20 °C)
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003377
CAS Number: 1122-91-4
EINECS: 214-365-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty