Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Adamantanone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Adamantan-2-one

A14275-14 A14275-06 A14275-14 A14275-06 A14275-22 A14275-22
AAA14275-06EA 50.59 USD InStock
AAA14275-06 AAA14275-14 CAAAA14275-06 AAA14275-22 CAAAA14275-14 CAAAA14275-22
2-Adamantanone 98%
2-Adamantanone


Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: ca 257 °C (subl.)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty