Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-bromopyridine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 5-Bromo-2-pyridylamine 5-Bromopyridin-2-amine

A14280-22 A14280-14 A14280-06 A14280-06 A14280-22 A14280-14
AAA14280-22EA 325.99 USD InStock
CAAAA14280-22 CAAAA14280-14 AAA14280-22 CAAAA14280-06 AAA14280-14 AAA14280-06
2-Amino-5-bromopyridine 98%
2-Amino-5-bromopyridine


Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 135…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty