Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 1-octansulfonate

A14292-22 A14292-14 A14292-14 A14292-06 A14292-22 A14292-06
AAA14292-22EA 714.29 USD InStock
AAA14292-22 AAA14292-14 CAAAA14292-06 CAAAA14292-14 AAA14292-06 CAAAA14292-22
1-Octanesulfonic acid sodium salt 99%
1-Octanesulphonic acid sodium salt
Reagent for HPLC analysis of peptides and proteins


Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty