Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitrovanillin 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde

A14296-14 A14296-06 A14296-22 A14296-06 A14296-22 A14296-14
AAA14296-14EA 114.99 USD InStock
CAAAAA14296-22 CAAAA14296-06 AAA14296-14 AAA14296-06 CAAAA14296-14 AAAA14296-22
5-Nitrovanillin 97%
5-Nitrovanillin


Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 174…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00007118
CAS Number: 6635-20-7
EINECS: 229-633-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty