Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexyltriphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14298-14 A14298-22 A14298-14 A14298-22 A14298-06 A14298-06
AAA14298-06EA 47.29 USD InStock
CAAAA14298-06 AAA14298-06 AAA14298-22 AAA14298-14 CAAAA14298-22 CAAAA14298-14
Cyclohexyltriphenylphosphonium bromide 98+%
Cyclohexyltriphenylphosphonium bromide


Formula: C₂₄H₂₆BrP
Melting Pt: 265…272 °C
Storage Temperature: Ambient
MDL Number: MFCD00031667
CAS Number: 7333-51-9
EINECS: 230-834-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty