Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Folic acid dihydrate 97%

Supplier: Thermo Scientific Chemicals

A14300-14 A14300-22 A14300-22 A14300-14
AAA14300-14EA 78.99 USD InStock
AAA14300-14 AAA14300-22 CAAAA14300-22 CAAAA14300-14
Folic acid dihydrate 97%
Folic acid dihydrate
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells


Formula: C₁₉H₁₉N₇O₆·2H₂O
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 75708-92-8
EINECS: 200-419-0
Order Now
Specification Test Results
Optical rotation +18° (c=0.5 in 0.1N NaOH)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty