Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Chloromethyl)trimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Danger

A14303-18 A14303-30 A14303-30 A14303-09 A14303-09 A14303-18
AAA14303-30EA 666.99 USD InStock
AAA14303-30 CAAAA14303-30 CAAAA14303-09 CAAAA14303-18 AAA14303-18 AAA14303-09
(Chloromethyl)trimethylsilane 98%
(Chloromethyl)trimethylsilane


Formula: ClH₂CSi(CH₃)₃
MW: 122.67 g/mol
Boiling Pt: 97…98 °C
Density: 0.886
Flash Pt: -2 °C(28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty