Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Methoxyphenyl)propionic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-(2-Methoxyphenyl)propanoic acid

A14314-22 A14314-06 A14314-22 A14314-06 A14314-14 A14314-14
AAA14314-06EA 36.99 USD InStock
CAAAA14314-22 AAA14314-06 AAA14314-14 CAAAA14314-14 AAA14314-22 CAAAA14314-06
3-(2-Methoxyphenyl)propionic acid 98+%
3-(2-Methoxyphenyl)propionic acid


Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002772
CAS Number: 6342-77-4
EINECS: 228-738-0
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty