Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N-methylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

A14343-06 A14343-14 A14343-06 A14343-03 A14343-03 A14343-14
AAA14343-14EA 488.99 USD InStock
CAAAA14343-06 AAA14343-14 AAA14343-03 CAAAA14343-03 CAAAA14343-14 AAA14343-06
2-Chloro-N-methylaniline 97%
2-Chloro-N-methylaniline


Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 218 °C
Density: 1.150
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045170
CAS Number: 932-32-1
EINECS: 000-000-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty