Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophenol 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Hydroxy-4-nitrobenzene

A14376-0I A14376-0I A14376-0B A14376-30 A14376-0B A14376-30
AAA14376-30EA 44.2 USD InStock
AAA14376-30 CAAAA14376-0B AAA14376-0I CAAAA14376-0I CAAAA14376-30 AAA14376-0B
4-Nitrophenol 99%
4-Nitrophenol


Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 279 °C (dec.)
Melting Pt: 110…115 °C
Density: 1.480
Flash Pt: 169 °C(336 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
EINECS: 202-811-7
UN: 1663
ADR: "6.1",III
Merck Index: 13,06654
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty