Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Azauracil 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

A14389-14 A14389-14 A14389-06 A14389-06
AAA14389-06EA 64.39 USD InStock
CAAAA14389-14 CAAAA14389-06 AAA14389-06 AAA14389-14
6-Azauracil 98%
6-Azauracil


Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 274…282 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty