Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14403-0B A14403-30 A14403-0I A14403-30 A14403-0B A14403-0I
AAA14403-0BEA 124.75 USD InStock
AAA14403-0B CAAAA14403-0I AAA14403-30 CAAAA14403-30 CAAAA14403-0B AAA14403-0I
Bromoacetic acid 98+%
Bromoacetic acid


Formula: BrCH₂COOH
MW: 138.95 g/mol
Boiling Pt: 207…209 °C
Melting Pt: 44…49 °C
Density: 1.930
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: "8",II
Merck Index: 13,01384
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty