Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Phosphonooxy)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Carboxyphenyl PhosphateFosfosal

A14417-14 A14417-14 A14417-06 A14417-06 A14417-03
AAA14417-06EA 402.99 USD InStock
AAA14417-06 AAAA14417-03 AAA14417-14 CAAAA14417-06 CAAAA14417-14
2-(Phosphonooxy)benzoic acid 98%
2-(Phosphonooxy)benzoic acid


Formula: C₇H₇O₆P
MW: 218.10 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00042645
CAS Number: 6064-83-1
EINECS: 227-993-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty