Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl nitroacetate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Nitroacetic ethyl ester

A14433-03 A14433-22 A14433-22 A14433-14 A14433-06 A14433-14 A14433-06 A14433-03
AAA14433-06EA 75.09 USD InStock
AAA14433-06 AAA14433-14 CAAAAA14433-22 CAAAA14433-06 AAAA14433-03 AAAA14433-22 CAAAAA14433-03 CAAAA14433-14
Ethyl nitroacetate 97%
Ethyl nitroacetate


Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 93…95 °C/10 mm
Density: 1,202
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty