Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Undecanol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-2-Hydroxy undecane (±)-Methyl nonyl carbinol (±)-Nonyl methyl carbinol

A14441-18 A14441-09 A14441-30 A14441-30 A14441-09 A14441-18
AAA14441-18EA 233.99 USD InStock
CAAAA14441-09 CAAAA14441-18 AAA14441-18 CAAAA14441-30 AAA14441-30 AAA14441-09
(±)-2-Undecanol 98+%
(±)-2-Undecanol


Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 228…229 °C
Melting Pt: 1…3 °C
Density: 0.828
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021958
CAS Number: 1653-30-1
EINECS: 216-722-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty