Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylmethanesulfonamide 98+%

Supplier: Thermo Scientific Chemicals

A14454-06 A14454-14 A14454-14 A14454-22 A14454-06 A14454-22
AAA14454-06EA 29.09 USD InStock
AAA14454-06 AAA14454-14 CAAAA14454-22 CAAAA14454-14 AAA14454-22 CAAAA14454-06
N,N-Dimethylmethanesulfonamide 98+%
N,N-Dimethylmethanesulphonamide


Formula: C₃H₉NO₂S
Boiling Pt: 119…120 °C/17 mm
Melting Pt: 46…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00014740
CAS Number: 918-05-8
EINECS: 213-039-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty