Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-pentylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Bromophenyl)pentane4-Bromopentylbenzene4-Pentylbromobenzene

A14457-03 A14457-06 A14457-03 A14457-06
AAA14457-06EA 107.99 USD InStock
AAA14457-06 CAAAA14457-06 AAA14457-03 CAAAA14457-03
1-Bromo-4-pentylbenzene 98%
1-Bromo-4-pentylbenzene


Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061113
CAS Number: 51554-95-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty