Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxyphenylboronic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (4-Methoxyphenyl)metaboric acid 4-Methoxybenzeneboronic acid Dihydroxy(4-methoxyphenyl)borane

A14462-14 A14462-03 A14462-06 A14462-03 A14462-14 A14462-06
AAA14462-14EA 365.99 USD InStock
CAAAA14462-03 CAAAA14462-14 AAA14462-14 AAA14462-03 AAA14462-06 CAAAA14462-06
4-Methoxyphenylboronic acid 97+%
4-Methoxyphenylboronic acid


Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 202…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty