Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Methylacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl p-tolyl ketone p-Methylacetophenone

A14469-18 A14469-30 A14469-18 A14469-0B A14469-0B A14469-30
AAA14469-30EA 39.19 USD InStock
CAAAA14469-30 AAA14469-30 CAAAA14469-0B AAA14469-18 AAA14469-0B CAAAA14469-18
4'-Methylacetophenone 96%
4'-Methylacetophenone


Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 224…226 °C
Melting Pt: –19 °C
Density: 1.010
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty