Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Furoylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Fur-2-yl)-3-oxopropanenitrile

A14497-06 A14497-03 A14497-03 A14497-14 A14497-06 A14497-14
AAA14497-06EA 107.53 USD InStock
CAAAA14497-06 CAAAA14497-03 CAAAA14497-14 AAA14497-06 AAA14497-03 AAA14497-14
2-Furoylacetonitrile 97%
2-Furoylacetonitrile


Formula: C₇H₅NO₂
MW: 135.12
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052210
CAS Number: 31909-58-7
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty