Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(4-Bromobutyl)phthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

A14517-06 A14517-06 A14517-22 A14517-14 A14517-22 A14517-14
AAA14517-22EA 782.99 USD InStock
AAA14517-22 AAA14517-14 CAAAA14517-06 CAAAA14517-14 AAA14517-06 CAAAA14517-22
N-(4-Bromobutyl)phthalimide 98%
N-(4-Bromobutyl)phthalimide


Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Boiling Pt: 165…170 °C/1 mm
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty