Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Cyclodextrine hydrate

Supplier: Thermo Scientific Chemicals

A14529-22 A14529-22 A14529-14 A14529-14 A14529-06 A14529-06
AAA14529-22EA 173.99 USD InStock
CAAAA14529-22 AAA14529-22 AAA14529-14 AAA14529-06 CAAAA14529-06 CAAAA14529-14
β-Cyclodextrine hydrate
β-Cyclodextrine hydrate


Formula: C₄₂H₇₀O₃₅·H₂O
Melting Pt: 288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150811
CAS Number: 68168-23-0
EINECS: 231-493-2
Order Now
Specification Test Results
Optical rotation +142° (c=1.5 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty