Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethyl methylenebisphosphonate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetraethyl methylenediphosphonateTetraethyl methylenebis(phosphonate)

A14532-22 A14532-14 A14532-06 A14532-06 A14532-22 A14532-14
AAA14532-22EA 1064.99 USD InStock
CAAAA14532-14 CAAAA14532-06 AAA14532-22 AAA14532-06 AAA14532-14 CAAAA14532-22
Tetraethyl methylenebisphosphonate 98+%
Tetraethyl methylenebisphosphonate


Formula: C₉H₂₂O₆P₂
MW: 288.22 g/mol
Boiling Pt: 151…153 °C/1.2 mm
Density: 1,162
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039887
CAS Number: 1660-94-2
EINECS: 216-764-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty