Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Cellobiose 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-O-β-D-Glucopyranosyl-D-glucopyranose β-CellobioseCellose4-β-D-glucopyranosyl-D-glueopyranoseβ-D-Glc-(1→4)-D-Glc

A14553-14 A14553-14 A14553-06 A14553-22 A14553-22 A14553-06
AAA14553-22EA 427.37 USD InStock
AAA14553-22 CAAAA14553-06 AAA14553-14 AAA14553-06 CAAAA14553-14 CAAAA14553-22
D-(+)-Cellobiose 98+%
D(+)-Cellobiose


Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 234 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5
Order Now
Specification Test Results
Optical rotation +34° (c=10 in water, 15h)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty