Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium-L(+)-tartrate hemihydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: (+)-Dipotassium L-tartrate hemihydrate (+)-Dipotassium-L-tartrat hemihydrate (R,R)-(+)-Dipotassium tartrate hemihydrate (R,R)-(+)-Potassium tartrate dibasic hemihydrate (R,R)-(+)-Tartaric acid dipotassium salt hemihydrate L(+)-Potassium tartrate dibasic hemihydrate L(+)-Tartaric acid dipotassium salt hemihydrate

A14556-0I A14556-0I A14556-30 A14556-0B A14556-0B A14556-30
AAA14556-30EA 64.04 USD InStock
CAAAA14556-0B AAA14556-30 AAA14556-0B CAAAA14556-30 CAAAA14556-0I AAA14556-0I
Dipotassium-L(+)-tartrate hemihydrate 99%
di-Potassium L(+)-tartrate hemihydrate


Formula: KOOC(CHOH)₂COOK·0,5H₂O
MW: 235.28 g/mol
Boiling Pt: 200…220 °C
Melting Pt: 155 °C (–H₂O of crystall.)
Density: 1.98
Storage Temperature: Ambient
MDL Number: MFCD00150136
CAS Number: 6100-19-2
EINECS: 213-067-8
Merck Index: 12,07849
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty