Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-1-hexanol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-6-hydroxyhexane6-Chlorohexan-1-ol6-Chlorohexanol

A14582-14 A14582-14 A14582-22 A14582-36 A14582-22 A14582-36
AAA14582-22EA 194 USD InStock
AAA14582-22 AAA14582-14 AAA14582-36 CAAAA14582-22 CAAAA14582-14 CAAAA14582-36
6-Chloro-1-hexanol 97%
6-Chloro-1-hexanol


Formula: C₆H₁₃ClO
MW: 136.62 g/mol
Boiling Pt: 109…111 °C/14 mm
Density: 1.027
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002984
CAS Number: 2009-83-8
EINECS: 217-925-2
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty