Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glutaric acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,5-Pentanedioic acid

A14595-14 A14595-36 A14595-22 A14595-14 A14595-36 A14595-22
AAA14595-22EA 80.89 USD InStock
AAA14595-22 AAA14595-14 CAAAA14595-36 CAAAA14595-14 AAA14595-36 CAAAA14595-22
Glutaric acid 99%
Glutaric acid


Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 200 °C/20 mm
Melting Pt: 95…99 °C
Density: 1,429
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty