Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-5-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Fluoro-2-methyl-4-nitrobenzene

A14649-09 A14649-09 A14649-18 A14649-18
AAA14649-18EA 188.99 USD InStock
CAAAA14649-09 CAAAA14649-18 AAA14649-18 AAA14649-09
2-Fluoro-5-nitrotoluene 98+%
2-Fluoro-5-nitrotoluene


Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 98…101 °C/13 mm
Melting Pt: 37…40 °C
Flash Pt: 105 °C(221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007284
CAS Number: 455-88-9
EINECS: 207-251-7
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty