Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Acetyl-1,2,3,4-tetrahydronaphthalene 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone

A14666-22 A14666-06 A14666-06 A14666-22 A14666-14 A14666-14
AAA14666-14EA 149.99 USD InStock
CAAAAA14666-22 CAAAA14666-06 CAAAA14666-14 AAA14666-14 AAAA14666-22 AAA14666-06
6-Acetyl-1,2,3,4-tetrahydronaphthalene 97%
6-Acetyl-1,2,3,4-tetrahydronaphthalene


Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 119…121 °C/2 mm
Melting Pt: –21 °C
Density: 1.058
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019710
CAS Number: 774-55-0
EINECS: 212-266-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty