Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Isopropylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

A14671-36 A14671-22 A14671-36 A14671-14 A14671-22 A14671-14
AAA14671-22EA 82.39 USD InStock
CAAAA14671-36 CAAAA14671-14 AAA14671-22 AAA14671-36 AAA14671-14 CAAAA14671-22
2-Isopropylaniline 97%
2-Isopropylaniline


Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 214…216 °C
Density: 0,966
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007720
CAS Number: 643-28-7
EINECS: 211-397-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty