Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-nitrothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-5-nitro-1,3-thiazole

A14701-03 A14701-06 A14701-14 A14701-14 A14701-06 A14701-03
AAA14701-06EA 96.19 USD InStock
CAAAA14701-06 AAA14701-06 AAA14701-14 AAA14701-03 CAAAA14701-03 CAAAA14701-14
2-Bromo-5-nitrothiazole 98%
2-Bromo-5-nitrothiazole


Formula: C₃HBrN₂O₂S
MW: 209.02 g/mol
Melting Pt: 85…90 °C
Density: 2,295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005317
CAS Number: 3034-48-8
EINECS: 221-226-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty