Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraphenylmethane 96%

Supplier: Thermo Scientific Chemicals
Warning

A14706-03 A14706-14 A14706-06 A14706-03 A14706-06 A14706-14
AAA14706-06EA 350.99 USD InStock
AAA14706-06 AAA14706-03 AAA14706-14 CAAAA14706-03 CAAAA14706-14 CAAAA14706-06
Tetraphenylmethane 96%
Tetraphenylmethane


Formula: C₂₅H₂₀
MW: 320.43 g/mol
Boiling Pt: 431 °C
Melting Pt: 280…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00014428
CAS Number: 630-76-2
EINECS: 211-144-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/63/2299863.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty