Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminocaproic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Aminohexanoic acid H-Eahx-OH ε-Amino-n-caproic acid

A14719-22 A14719-0E A14719-0E A14719-36 A14719-22 A14719-36
AAA14719-36EA 196.99 USD InStock
CAAAA14719-36 CAAAA14719-22 AAA14719-36 CAAAA14719-0E AAA14719-0E AAA14719-22
6-Aminocaproic acid 99%
6-Aminocaproic acid


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00008238
CAS Number: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty