Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-α,α,α-trifluorotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-3-(trifluoromethyl)benzene 3-Bromobenzotrifluoride

A14724-14 A14724-36 A14724-14 A14724-36 A14724-22 A14724-22
AAA14724-22EA 110.99 USD InStock
AAA14724-22 AAA14724-14 CAAAA14724-14 CAAAA14724-22 AAAA14724-36 CAAAAA14724-36
3-Bromo-α,α,α-trifluorotoluene 98+%
3-Bromo-α,α,α-trifluorotoluene


Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 151…152 °C
Melting Pt: 37.5…39 °C
Density: 1.613
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000380
CAS Number: 401-78-5
EINECS: 206-932-6
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty