Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Arginine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-(+)-Arginine hydrochloride H-L-Arg-OH.HCl H-Arg-OH.HCl

A14730-22 A14730-22 A14730-0E A14730-0E A14730-36 A14730-36
AAA14730-36EA 205.66 USD InStock
AAA14730-36 CAAAA14730-22 AAA14730-0E CAAAA14730-36 AAA14730-22 CAAAA14730-0E
L-Arginine hydrochloride 98+%
L-Arginine hydrochloride


Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 216 °C (dec.)
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty