Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl propiolate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propiolic acid methyl ester 2-Propynoic acid methyl ester

A14743-22 A14743-06 A14743-14 A14743-06 A14743-22 A14743-14
AAA14743-06EA 59.69 USD InStock
CAAAA14743-22 AAA14743-06 CAAAA14743-14 CAAAA14743-06 AAA14743-14 AAA14743-22
Methyl propiolate 99%
Methyl propiolate


Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 98…101 °C
Density: 1.016
Flash Pt: 10 °C(50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008572
CAS Number: 922-67-8
EINECS: 213-083-5
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty