Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Picolinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pyridine-2-carboxylic acid 2-Pyridinecarboxylic acid

A14749-22 A14749-36 A14749-36 A14749-22
AAA14749-22EA 70.72 USD InStock
AAA14749-22 CAAAA14749-36 CAAAA14749-22 AAA14749-36
Picolinic acid 99%
Picolinic acid


Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 135…139 °C
Density: 1,49 g/cm³ (20 °C)
Flash Pt: 139 °C(232 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006293
CAS Number: 98-98-6
EINECS: 202-719-7
Merck Index: 12,07557
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty