Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methyl-2-pyridone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Hydroxy-6-methylpyridine 6-Methylpyridin-2-ol

A14764-14 A14764-06 A14764-06 A14764-14
AAA14764-06EA 48.49 USD InStock
CAAAA14764-06 AAA14764-06 CAAAA14764-14 AAA14764-14
6-Methyl-2-pyridone 98%
6-Methyl-2-pyridone


Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 157…161 °C
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty