Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Mandelic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D(-)-2-Hydroxy-2-phenylacetic acid (R)-(-)-2-Hydroxy-2-phenylacetic acid D(-)-Mandelic acid

A14767-22 A14767-06 A14767-22 A14767-14 A14767-06 A14767-14
AAA14767-22EA 228.99 USD InStock
CAAAA14767-06 CAAAA14767-14 AAA14767-22 CAAAA14767-22 AAA14767-06 AAA14767-14
(R)-(-)-Mandelic acid 98%
(R)-(-)-Mandelic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130…134 °C
Density: 1,341 g/cm³ (20 °C)
Flash Pt: >190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty