Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Methylthio)propanal 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-(Methylthio)propionaldehyde

A14774-18 A14774-30 A14774-18 A14774-09 A14774-30 A14774-09
AAA14774-30EA 494.99 USD InStock
CAAAA14774-18 AAA14774-30 AAA14774-09 CAAAA14774-30 AAA14774-18 CAAAA14774-09
3-(Methylthio)propanal 98%
3-(Methylthio)propanal


Formula: C₄H₈OS
MW: 104.17 g/mol
Boiling Pt: 165…166 °C
Melting Pt: –58 °C
Density: 1.052
Flash Pt: 64 °C(147 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007022
CAS Number: 3268-49-3
EINECS: 221-882-5
UN: 2785
ADR: "6.1",III
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty