Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitrocoumarin 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14792-22 A14792-06 A14792-14 A14792-22 A14792-06 A14792-14
AAA14792-22EA 582.75 USD InStock
CAAAA14792-06 CAAAA14792-14 AAA14792-22 CAAAA14792-22 AAA14792-06 AAA14792-14
6-Nitrocoumarin 98+%
6-Nitrocoumarin


Formula: C₉H₅NO₄
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00016973
CAS Number: 2725-81-7
EINECS: 220-341-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty