Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Acetylnaphthalene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2'-Acetonaphthone Methyl β-naphthyl ketone

A14793-18 A14793-30 A14793-22 A14793-0B A14793-18 A14793-30 A14793-22 A14793-0B
AAA14793-30EA 83.89 USD InStock
CAAAA14793-0B AAA14793-30 AAA14793-0B CAAAAA14793-22 CAAAA14793-18 AAAA14793-22 AAA14793-18 CAAAA14793-30
2-Acetylnaphthalene 99%
2-Acetylnaphthalene


Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300…301 °C
Melting Pt: 52…56 °C
Density: 1.1171 g/cm³ (20 °C)
Flash Pt: 168 °C(334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004108
CAS Number: 93-08-3
EINECS: 202-216-2
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty