Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenyl vinyl sulfone 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (Vinylsulphonyl)benzene Vinyl phenyl sulphone

A14794-14 A14794-06 A14794-06 A14794-14 A14794-03 A14794-03
AAA14794-14EA 358.99 USD InStock
CAAAA14794-14 CAAAA14794-03 CAAAA14794-06 AAA14794-14 AAA14794-03 AAA14794-06
Phenyl vinyl sulfone 99+%
Phenyl vinyl sulphone


Formula: C₈H₈O₂S
MW: 168.22 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007554
CAS Number: 5535-48-8
EINECS: 226-890-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty