Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Acetylvaleric acid 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 6-Oxoheptanoic acid

A14810-18 A14810-06 A14810-18 A14810-09 A14810-09 A14810-06
AAAA14810-06EA 82.19 USD InStock
CAAAA14810-18 CAAAAA14810-06 AAAA14810-06 CAAAA14810-09 AAA14810-18 AAA14810-09
5-Acetylvaleric acid 96%
5-Acetylvaleric acid
Soluble in water


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 138…140 °C/1.5 mm
Melting Pt: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00051479
CAS Number: 3128-07-2
EINECS: 221-512-2
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty