Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(Ethoxycarbonyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl 1,3-dioxoisoindoline 2-carboxylateN-Carbethoxyphthalimide

A14822-36 A14822-14 A14822-36 A14822-22 A14822-22 A14822-14
AAA14822-14EA 41.49 USD InStock
AAA14822-14 AAA14822-36 AAA14822-22 CAAAA14822-22 CAAAA14822-36 CAAAA14822-14
N-(Ethoxycarbonyl)phthalimide 97%
N-(Ethoxycarbonyl)phthalimide


Formula: C₁₁H₉NO₄
MW: 219.20 g/mol
Melting Pt: 78…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005893
CAS Number: 22509-74-6
EINECS: 245-048-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty