Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Elaidic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (E)-Octadec-9-enoic acid

A14832-14 A14832-03 A14832-14 A14832-03 A14832-06 A14832-06
AAA14832-14EA 774.99 USD InStock
CAAAA14832-06 AAA14832-14 AAA14832-03 AAA14832-06 CAAAA14832-14 CAAAA14832-03
Elaidic acid 98%
Elaidic acid


Formula: C₁₈H₃₄O₂
MW: 282.47 g/mol
Boiling Pt: 288 °C/100 mm
Melting Pt: 41…45 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063954
CAS Number: 112-79-8
EINECS: 204-006-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty